missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, MP Biomedicals™

Product Code. 11491771 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491771 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11491771 Supplier MP Biomedicals Supplier No. 02190271.5
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used for a variety of applications. Sucrose, MP Biomedicals is a simple carbohydrate isolated from cane sugar; it is not synthetic. Sucrose is a disaccharide composed of glucose and fructose that can be used to prepare density gradients, culture media and

Sucrose can be used to prepare density gradients for cell/organelle separation. The refractive index of the solution will be directly related to the density. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays.

Specifications

CAS 57-50-1
Absorbance A50%/260 ≤0.15; A50%/280 ≤0.15 (Lit.)
Melting Point 177°C to 187°C
Density 1.587 at 25°C (water=1) (Lit.)
Molecular Formula C12H22O11
MDL Number MFCD00006626
Quantity 500 g
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
Solubility Information Soluble in water (1g/0.5mL at 25°C; 1g/0.2mL boiling water), ethanol (1g/170mL) or methanol (1g/100mL); moderately soluble in glycerol or pyridine.
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Optical Rotation 66.0° ± 5 (H2O, NH4OH)
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
PubChem CID 5988
ChEBI CHEBI:17992
Formula Weight 342.30
Percent Purity ≥95%
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.