missing translation for 'onlineSavingsMsg'
Learn More

Sucrose Octasulfate, Ammonium Salt, TRC

Product Code. 30424259
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424259 5 g 5g
30425743 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424259 Supplier Toronto Research Chemicals Supplier No. TRCS6989905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sucrose Octasulfate','Ammonium Salt
CAS 74135-13-0
Purity Grade Notes HPLC
Molecular Formula C12 H14 O35 S8 . 8 H4 N
Quantity 5 g
Synonym α-D-Glucopyranoside','1,3,4,6-tetra-O-sulfo-β-D-fructofuranosyl','tetrakis(hydrogen sulfate)','octaammonium salt (9CI), 1,3,4,6-Tetra-O-sulfo-β-D-fructofuranosyl-α-D-glucopyranoside 2,3,4,6-tetrakis(hydrogen sulfate) ammonium salt (1:8), Sucrosofate ammonium, Ammonium sucrose octasulfate, Sucrose octasulfate ammonium salt, 1,3,4,6-Tetra-O-sulfo-β-D-fructofuranosyl-α-D-glucopyranoside tetrakis(hydrogen sulfate) octaammonium salt
InChI Formula InChI=1S/C12H22O35S8.8H3N/c13-48(14,15)37-1-4-6(43-51(22,23)24)8(45-53(28,29)30)9(46-54(31,32)33)11(40-4)42-12(3-39-50(19,20)21)10(47-55(34,35)36)7(44-52(25,26)27)5(41-12)2-38-49(16,17)18;;;;;;;;/h4-11H,1-3H2,(H,13,14,15)(H,16,17,18)(H,19,20,21)(H,22,23,24)(H,25,26,27)(H,28,29,30)(H,31,32,33)(H,34,35,36);8*1H3/t4-,5-,6-,7-,8+,9-,10+,11-,12+;;;;;;;;/m1......../s1
SMILES [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[O-]S(=O)(=O)OC[C@H]1O[C@H](O[C@]2(COS(=O)(=O)[O-])O[C@H](COS(=O)(=O)[O-])[C@@H](OS(=O)(=O)[O-])[C@@H]2OS(=O)(=O)[O-])[C@H](OS(=O)(=O)[O-])[C@@H](OS(=O)(=O)[O-])[C@@H]1OS(=O)(=O)[O-]
IUPAC Name octaazanium;[(2R,3R,4S,5R,6R)-2-[(2S,3S,4R,5R)-3,4-disulfonatooxy-2,5-bis(sulfonatooxymethyl)oxolan-2-yl]oxy-3,5-disulfonatooxy-6-(sulfonatooxymethyl)oxan-4-yl] sulfate
Molecular Weight (g/mol) 1119.05
Formula Weight 1117.9831
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.