missing translation for 'onlineSavingsMsg'
Learn More

Sucrose Octasulfate Octatriethylamine Salt (>90%), TRC

Product Code. 30423766
Change view
Click to view available options
Quantity:
500 mg
2.5 g
5 g
Unit Size:
2.5g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423766 500 mg 500mg
30414529 2.5 g 2.5g
30412408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423766 Supplier Toronto Research Chemicals Supplier No. TRCS698992500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869561-07-9
Molecular Formula C12H22O35S8 . 8(C6H15N)
Molecular Weight (g/mol) 1792.3
Synonym Sucrosefate Triethylamine Salt, 1,3,4,6-Tetra-O-sulfo-beta-D-fructofuranosyl alpha-D-Glucopyranoside Tetrakis(hydrogen Sulfate) TEA Salt
IUPAC Name N,N-diethylethanamine;[(2R,3R,4S,5R,6R)-2-[(2S,3S,4R,5R)-3,4-disulfooxy-2,5-bis(sulfooxymethyl)oxolan-2-yl]oxy-3,5-disulfooxy-6-(sulfooxymethyl)oxan-4-yl] hydrogen sulfate
SMILES CCN(CC)CC.OS(=O)(=O)OC[C@H]1O[C@H](O[C@]2(COS(=O)(=O)O)O[C@H](COS(=O)(=O)O)[C@@H](OS(=O)(=O)O)[C@@H]2OS(=O)(=O)O)[C@H](OS(=O)(=O)O)[C@@H](OS(=O)(=O)O)[C@@H]1OS(=O)(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H22O35S8.C6H15N/c13-48(14,15)37-1-4-6(43-51(22,23)24)8(45-53(28,29)30)9(46-54(31,32)33)11(40-4)42-12(3-39-50(19,20)21)10(47-55(34,35)36)7(44-52(25,26)27)5(41-12)2-38-49(16,17)18;1-4-7(5-2)6-3/h4-11H,1-3H2,(H,13,14,15)(H,16,17,18)(H,19,20,21)(H,22,23,24)(H,25,26,27)(H,28,29,30)(H,31,32,33)(H,34,35,36);4-6H2,1-3H3/t4-,5-,6-,7-,8+,9-,10+,11-,12+;/m1./s1
Formula Weight 1082.8912
Percent Purity >95%
Chemical Name or Material Sucrose Octasulfate Octatriethylamine Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.