missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sucrose palmitate, 90%

Product Code. 15458059
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
Product Code. Quantity unitSize
15458059 50 g 50g
15468059 250 g 250g
15478059 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 15458059

Brand: Thermo Scientific Chemicals J66627.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 26446-38-8 | C28H52O12

Sucrose palmitate is used as food emulsifier. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose palmitate is used as food emulsifier. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in DMSO, DMF, ethanol and water

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26446-38-8
Molecular Formula C28H52O12
MDL Number MFCD00047545
Synonym beta-D-Fructofuranosyl-alpha-D-glucopyranoside monohexadecanoate

Specifications

Color White
Quantity 50 g
Solubility Information Soluble in DMSO, DMF, ethanol and water
Formula Weight 508.72
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Sucrose palmitate
compliance-icons
Product Identifier
  • Sucrose palmitate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sucrose palmitate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.