missing translation for 'onlineSavingsMsg'
Learn More

Sucrose Stearate - 70% monostearate, TRC

Product Code. 30012442
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012442 5 g 5g
30010167 50 g 50g
30011230 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012442 Supplier Toronto Research Chemicals Supplier No. TRCS6990385G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25168-73-4
Molecular Formula C30 H56 O12
Molecular Weight (g/mol) 608.76
Synonym alpha-D-Glucopyranoside beta-D-fructofuranosyl monooctadecanoate, Sucrose monostearate, Stearic acid monoester with sucrose, NSC 192745, Ryoto S 1670, Ryoto Sugar Ester MO-C 18, Ryoto Sugar Ester S 1670, S 1670, Saccharose monostearate, Stearic acid sucrose ester, Sucrapan S, Sucrose monooctadecanoate, Sucrose monostearic acid ester, Sugar Ester S 1670, Surfhope SE Pharma J-1809
IUPAC Name [(2S,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl]oxy-tetrahydrofuran-2-yl]methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC[C@@]1(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Quantity 5 g
Chemical Name or Material alpha-D-Glucopyranoside beta-D-fructofuranosyl monooctadecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.