missing translation for 'onlineSavingsMsg'
Learn More

Sudan Black, Fisher BioReagents™

Product Code. 10491135
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10491135 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10491135 Supplier Fisher BioReagents Supplier No. BP10910

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sudan Black is used to stain chromosomes and lipoproteins, as well as fat in bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 120°C
CAS 4197-25-5
Extinction Coefficient 600g(-1) cm(-1) min.
Molecular Formula C29H24N6
Merck Index 15, 9016
Identification Pass Test
InChI Key YCUVUDODLRLVIC-UHFFFAOYSA-N
SMILES CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C
IUPAC Name (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
Molecular Weight (g/mol) 456.553
PubChem CID 61336
Lambda Max 598 to 602nm
Formula Weight 456.54
Loss on Drying 5% max.
Packaging Glass Bottle
Color Index No. 26150
Color Brown
Quantity 10 g
Physical Form Powder
Chemical Name or Material Sudan Black
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.