missing translation for 'onlineSavingsMsg'
Learn More

Sudan III, C.I.26100, Fisher Chemical

Product Code. 12897182 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12897182 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12897182 Supplier Fisher Chemical Supplier No. S/8620/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

25g, Glass Jar

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 199°C
CAS 85-86-9
Molecular Formula C22H16N4O
MDL Number 3905
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
Molecular Weight (g/mol) 352.397
PubChem CID 6789251
Formula Weight 352.1382g/mol
Packaging Glass Jar
Color Brown/Red
Quantity 25 g
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.