missing translation for 'onlineSavingsMsg'
Learn More

Sudan III, TRC

Product Code. 30414467
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414467 25 g
30426976 10 g
30413232 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30414467 Supplier Toronto Research Chemicals Supplier No. TRCS68890025G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-86-9
Molecular Formula C22 H16 N4 O
Molecular Weight (g/mol) 352.39
Synonym 2-Naphthalenol, 1-[2-[4-(2-phenyldiazenyl)phenyl]diazenyl]- (ACI), 1-[2-[4-(2-Phenyldiazenyl)phenyl]diazenyl]-2-naphthalenol (ACI), 2-Naphthalenol, 1-[[4-(phenylazo)phenyl]azo]- (9CI), 2-Naphthol, 1-(p-phenylazophenylazo)- (6CI), C.I. Solvent Red 23 (8CI), 1-(p-Phenylazophenylazo)-2-naphthol, 1-(p-Phenylazophenylazo)-2-naphthol (Sudan III), 111440 Red, Brasilazina Oil Scarlet, C.I. 26100, Certiqual Oil Red, D and C Red No. 17, D&C Red 17, D&C Red 17 K 7007, DC Red 17, Fast Oil Scarlet III, Fat Red HRR, Fat Red R, Fat Red RS, Fat Scarlet LB, Fat Soluble Red Zh, FD And C Red No. 17, Grasal Brilliant Red G, Grasan Brilliant Red G, Japan Red 225, Japan Red No. 225, NSC 65825, NSC 8995, Oil Red 3G, Oil Red AS, Oil Red DR 126, Oil Red Extra, Oil Scarlet G, Organol Red BS, Organol Scarlet, Red 17, Red No. 225, Red Zh, Silotras Scarlet TB, Solvent Red 23, Somalia Red III, Stearix Scarlet, Sudan 3, Sudan III, Sudan P III, Sudan Red III, Tetrazobenzene-β-naphthol, Toney Red, VisioTag
IUPAC Name 1-[(E)-[4-[(E)-phenyldiazenyl]phenyl]diazenyl]naphthalen-2-ol
SMILES Oc1ccc2ccccc2c1N=Nc3ccc(cc3)N=Nc4ccccc4

Specifications

Quantity 25 g
InChI Formula InChI=1S/C22H16N4O/c27-21-15-10-16-6-4-5-9-20(16)22(21)26-25-19-13-11-18(12-14-19)24-23-17-7-2-1-3-8-17/h1-15,27H
Formula Weight 352.1324
Chemical Name or Material Sudan III
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.