missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan IV

Product Code. 11444823
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444823 25 g 25g
11454823 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444823 Supplier Thermo Scientific Chemicals Supplier No. A12181.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 85-83-6 | C24H20N4O | 380.45 g/mol

Useful as a stain for fat in animal tissue

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-83-6
Molecular Formula C24H20N4O
Molecular Weight (g/mol) 380.45
MDL Number MFCD00003893
InChI Key KMDLOETUWUPGMB-BXCCFQQFSA-N
Synonym C.I. 26105; Scarlet Red
PubChem CID 6797604
IUPAC Name (1Z)-1-(2-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
SMILES CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C

Specifications

Melting Point ∼185°C (decomposition)
Merck Index 14,8393
Formula Weight 380.45
Quantity 25 g
Chemical Name or Material Sudan IV
compliance-icons
Product Identifier
  • Sudan IV
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sudan IV

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.