missing translation for 'onlineSavingsMsg'
Learn More

Sudan IV (>80%), TRC

Product Code. 30421287
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421287 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30421287 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCS68892050MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sudan IV
CAS 85-83-6
Molecular Formula C24 H20 N4 O
Quantity 50 mg
Synonym 2-Naphthalenol','1-[[2-methyl-4-[(2-methylphenyl)azo]phenyl]azo]- (9CI), 2-Naphthol','1-(4-o-tolylazo-o-tolylazo)- (6CI), C.I. Solvent Red 24 (8CI), 1-[2-[2-Methyl-4-[2-(2-methylphenyl)diazenyl]phenyl]diazenyl]-2-naphthalenol, 1-(2'-Methyl-4'-(2''-methylphenylazo)phenylazo)-2-naphthol, 1-[4-(o-Tolylazo)-o-tolylazo]-2-naphthol, 2',3-Dimethyl-4-(2-hydroxynaphthylazo)azobenzene, Aizen SOT Red 1, Atlasol Red 4B, Biebrich Scarlet BPC, Biebrich Scarlet Red, Brasilazina Oil Red B, C.I. 26105, Candle Scarlet 2B, Candle Scarlet B, Candle Scarlet G, Ceres Red BB, Dispersol Red PP, Enial Red IV, Fast Oil Red B, Fast Red BB, Fat Ponceau R, Fat Red 2B, Fat Red B, Fat Red BB, Fat Red BS, Fat Red TS, Fat Soluble Dark Red, Grasal Brilliant Red B, Grasan Brilliant Red B, Hidaco Oil Red, Japan Red 501, Lacquer Red V, Lacquer Red VS, Medical Scarlet, NSC 10472, Oil Red, Oil Red 282, Oil Red 2B, Oil Red 3, Oil Red 3B, Oil Red 47, Oil Red 7, Oil Red A, Oil Red APT, Oil Red B, Oil Red BB, Oil Red BS, Oil Red D, Oil Red ED, Oil Red F, Oil Red GO, Oil Red RC, Oil Red RR, Oil Red S, Oil Red SST Extra, Oil Red TAX, Oil Red ZD, Oil Scarlet, Oil Scarlet 48, Oleal Red BB, Oleosol Red BB, Orcosolve Red 4B, Organol Red B, Orient Oil Red RR, Permalex Red 2B, Plastoresin Red F, Red 3R Soluble in Grease, Red No. 501, Resinol Red 2B, SOT Red 1, Scarlet R, Scarlet R (solvent dye), Scarlet oil, Scarlet red, Silotras Red T 3B, Solvent Red 24, Somalia Red IV, Stearix Red 4B, Stearix Red 4S, Sudan (IV) dye, Sudan 4, Sudan IV, Sudan P, Sudan Red 380, Sudan Red 4BA, Sudan Red BB, Sudan Red BBA, Sudan Red IV, TU 6-14-8780, Tertrogras Red N, Toyo Oil Red BB, Waxoline Red O, Waxoline Red OM, Waxoline Red OS, o-Tolueneazo-o-toluene-β-naphthol, o-Tolylazo-o-tolylazo-2-naphthol, o-Tolylazo-o-tolylazo-β-naphthol
InChI Formula InChI=1S/C24H20N4O/c1-16-7-3-6-10-21(16)26-25-19-12-13-22(17(2)15-19)27-28-24-20-9-5-4-8-18(20)11-14-23(24)29/h3-15,29H,1-2H3
SMILES Cc1ccccc1N=Nc2ccc(N=Nc3c(O)ccc4ccccc34)c(C)c2
IUPAC Name 1-[(E)-[2-methyl-4-[(E)-(2-methylphenyl)diazenyl]phenyl]diazenyl]naphthalen-2-ol
Molecular Weight (g/mol) 380.44
Formula Weight 380.1637
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.