missing translation for 'onlineSavingsMsg'
Learn More

Sudan IV, MP Biomedicals™

Product Code. 11456115 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456115 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11456115 Supplier MP Biomedicals Supplier No. 0215264825
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 85-83-6
Molecular Formula C24H20N4O
MDL Number MFCD00003893
Synonym Fat ponceau, Lipid crimson
InChI Key KMDLOETUWUPGMB-BXCCFQQFSA-N
SMILES CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C
IUPAC Name 1-[[2-methyl-4-[(2-methylphenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-one
Molecular Weight (g/mol) 380.45
PubChem CID 6797604
Formula Weight 380.4
Color Brown to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Sudan IV
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.