missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan Red 7B, TRC
Specifications
Specifications
| Chemical Name or Material | Sudan Red 7B |
| CAS | 6368-72-5 |
| Molecular Formula | C24 H21 N5 |
| Quantity | 500 mg |
| Synonym | 2-Naphthalenamine','N-ethyl-1-[2-[4-(2-phenyldiazenyl)phenyl]diazenyl]- (ACI), 2-Naphthalenamine','N-ethyl-1-[[4-(phenylazo)phenyl]azo]- (9CI), C.I. Solvent Red 19 (7CI','8CI), N-Ethyl-1-[2-[4-(2-phenyldiazenyl)phenyl]diazenyl]-2-naphthalenamine (ACI), Sudan Red 7B (6CI), (Phenylazo-4-phenylazo)-1-ethylamino-2-naphthalene, C.I. 26050, Ceres Red 7B, Fat Red 7B, Fat red bluish, Hexatype Carmine B, Lacquer Red V 3B, NSC 326202, Oil Violet, Organol Bordeaux B, Solvent Red 19, Special Blue X 2137, Sudan 7B, Sudan Red 460, Sudan Red 462, Typogen Carmine |
| InChI Formula | InChI=1S/C24H21N5/c1-2-25-23-17-12-18-8-6-7-11-22(18)24(23)29-28-21-15-13-20(14-16-21)27-26-19-9-4-3-5-10-19/h3-17,25H,2H2,1H3 |
| SMILES | CCNc1ccc2ccccc2c1N=Nc3ccc(cc3)N=Nc4ccccc4 |
| IUPAC Name | N-ethyl-1-[(E)-[4-[(E)-phenyldiazenyl]phenyl]diazenyl]naphthalen-2-amine |
| Molecular Weight (g/mol) | 379.46 |
| Formula Weight | 379.1797 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?