missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan Red 7B, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Sudan Red 7B |
| CAS | 6368-72-5 |
| Molecular Formula | C24 H21 N5 |
| Quantity | 1 g |
| Synonym | 2-Naphthalenamine','N-ethyl-1-[2-[4-(2-phenyldiazenyl)phenyl]diazenyl]- (ACI), 2-Naphthalenamine','N-ethyl-1-[[4-(phenylazo)phenyl]azo]- (9CI), C.I. Solvent Red 19 (7CI','8CI), N-Ethyl-1-[2-[4-(2-phenyldiazenyl)phenyl]diazenyl]-2-naphthalenamine (ACI), Sudan Red 7B (6CI), (Phenylazo-4-phenylazo)-1-ethylamino-2-naphthalene, C.I. 26050, Ceres Red 7B, Fat Red 7B, Fat red bluish, Hexatype Carmine B, Lacquer Red V 3B, NSC 326202, Oil Violet, Organol Bordeaux B, Solvent Red 19, Special Blue X 2137, Sudan 7B, Sudan Red 460, Sudan Red 462, Typogen Carmine |
| InChI Formula | InChI=1S/C24H21N5/c1-2-25-23-17-12-18-8-6-7-11-22(18)24(23)29-28-21-15-13-20(14-16-21)27-26-19-9-4-3-5-10-19/h3-17,25H,2H2,1H3 |
| SMILES | CCNc1ccc2ccccc2c1N=Nc3ccc(cc3)N=Nc4ccccc4 |
| IUPAC Name | N-ethyl-1-[(E)-[4-[(E)-phenyldiazenyl]phenyl]diazenyl]naphthalen-2-amine |
| Molecular Weight (g/mol) | 379.46 |
| Formula Weight | 379.1797 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction