missing translation for 'onlineSavingsMsg'
Learn More

Sulbactam, TRC

Product Code. 30137138
Change view
Click to view available options
Quantity:
50 mg
1 g
25 g
Unit Size:
1g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137138 1 g 1g
30134988 25 g 25g
30007031 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137138 Supplier Toronto Research Chemicals Supplier No. TRCS6889701G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68373-14-8
Molecular Formula C8 H11 N O5 S
Molecular Weight (g/mol) 233.24
Synonym Sulbactam, (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-Dioxide, (2S-cis)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-Dioxide, Betamaze, CP 45899, Penicillanic acid 1,1-dioxide, Penicillanic acid S,S-dioxide, Penicillanic acid dioxide, Penicillanic acid sulfone
IUPAC Name (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H11NO5S/c1-8(2)6(7(11)12)9-4(10)3-5(9)15(8,13)14/h5-6H,3H2,1-2H3,(H,11,12)/t5-,6+/m1/s1
Formula Weight 233.0358
Percent Purity >95%
Chemical Name or Material Sulbactam
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.