missing translation for 'onlineSavingsMsg'
Learn More

Sulclamide, TRC

Product Code. 30426978
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426978 10 g -
30416932 1 g -
30412009 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426978 Supplier Toronto Research Chemicals Supplier No. TRCS69901310G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2455-92-7
Molecular Formula C7H7ClN2O3S
Molecular Weight (g/mol) 234.66
Synonym 3-(Aminosulfonyl)-4-chlorobenzamide, 3-Sulfamido-4-chlorobenzamide, 3-Sulfamoyl-4-chlorobenzamide, 3-Sulfamyl-4-chlorobenzamide, 4-Chloro-3-sulfamoylbenzamide, SD 141-12, Sulclamid
IUPAC Name 4-chloro-3-sulfamoylbenzamide
SMILES NC(=O)c1ccc(Cl)c(c1)S(=O)(=O)N

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H7ClN2O3S/c8-5-2-1-4(7(9)11)3-6(5)14(10,12)13/h1-3H,(H2,9,11)(H2,10,12,13)
Formula Weight 233.9866
Chemical Name or Material Sulclamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.