missing translation for 'onlineSavingsMsg'
Learn More

Sulfadiazine Silver Salt, TRC

Product Code. 30240867
Change view
Click to view available options
Quantity:
10 mg
10 g
25 g
Unit Size:
10g
10mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240867 10 mg 10mg
30240756 25 g 25g
30240602 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240867 Supplier Toronto Research Chemicals Supplier No. TRCS69905010MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22199-08-2
Molecular Formula C10 H9 N4 O2 S . Ag
Molecular Weight (g/mol) 357.14
Synonym Silver, [4-amino-N-(2-pyrimidinyl-κN1)benzenesulfonamidato-κO]-, Silver, (4-amino-N-2-pyrimidinylbenzenesulfonamidato-NN,O1)-, Sulfanilamide, N1-2-pyrimidinyl-, monosilver(1+) salt (8CI), Benzenesulfonamide, 4-amino-N-2-pyrimidinyl-, silver complex, Altreet-Ag, Dermazin, Dermazine, Flamazine, Flammazine, Geben, NSC 625324, Physiotulle-Ag, Silvadene, Silver 2-sulfanilamidopyrimidine, Silver sulfadiazine, Silver sulphadiazine, Silverol, Sulfadiazine silver, Sulfadiazine silver salt
IUPAC Name silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide
SMILES [Ag+].Nc1ccc(cc1)S(=O)(=O)[N-]c2ncccn2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H9N4O2S.Ag/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10;/h1-7H,11H2;/q-1;+1
Formula Weight 355.9497
Chemical Name or Material Silver Sulfadiazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.