missing translation for 'onlineSavingsMsg'
Learn More

Sulfadiazine, TRC

Product Code. 17933025
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933025 50 mg 50mg
17921305 10 g 10g
17934955 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933025 Supplier Toronto Research Chemicals Supplier No. TRCS69905450MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-35-9
Molecular Formula C10 H10 N4 O2 S
Molecular Weight (g/mol) 250.28
Synonym 4-Amino-N-pyrimidin-2-ylbenzenesulfonamide, Sulfadiazine, Sulfadimidine Imp. B (Pharmeuropa), N1-2(1H)-Pyrimidinylidenesulfanilamide, N1-2-Pyrimidinylsulfanilamide, 2-(4-Aminobenzenesulfonamido)pyrimidine, 2-(4-Aminobenzenesulfonylamino)pyrimidine, 2-(p-Aminobenzenesulfonamido)pyrimidin, 2-Sulfanilamidopyrimidine, 2-Sulfanilylaminopyrimidine, 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide, 4-Amino-N-(2-pyrimidyl)benzenesulfonamide, 4-[[(Pyrimidin-2-yl)amino]sulfonyl]aniline, A 306, Adiazin, Adiazine, Coco-Diazine, Cremodiazine, Debenal, Deltazina, Di-Azo-Mul, Diazin, Diazolone, Diazovit, Diazyl, Eskadiazine, Honey diazine, Lipo-Diazine, Lipo-Levazine, Liquadiazine, Microsulfon, N1-2-Pyrimidinylsulfanilamide, N1-2-Pyrimidylsulfanilamide, NSC 35600, Neazine, Piridisir, Pirimal, Pyrimal, RP 2616, SDA, SN 112, Sanodiazine, Spofadrizine, Sterazine, Sulfadiazin, Sulfadiazina Reig Jofre, Sulfapirimidin, Sulfapyrimidine, Sulfazin, Sulfazin (Russian pharmaceutical), Sulfazine, Sulfolex, Sulphadiazine, Sulphadiazine E, Theradiazine
IUPAC Name 4-amino-N-pyrimidin-2-ylbenzenesulfonamide
SMILES Nc1ccc(cc1)S(=O)(=O)Nc2ncccn2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H10N4O2S/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10/h1-7H,11H2,(H,12,13,14)
Formula Weight 250.0524
Percent Purity >95%
Chemical Name or Material Sulfadiazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.