missing translation for 'onlineSavingsMsg'
Learn More

Sulfadoxine, 98%

Product Code. 15467559
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15467559 100 g Plastic bottle 100g
15447559 5 g Glass bottle 5g
15457559 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15467559 Supplier Thermo Scientific Alfa Aesar Supplier No. J66268.22
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2447-57-6 | C12H14N4O4S | 310.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2447-57-6
Molecular Formula C12H14N4O4S
Molecular Weight (g/mol) 310.33
MDL Number MFCD00792890
InChI Key PJSFRIWCGOHTNF-UHFFFAOYSA-N
Synonym sulfadoxine, sulforthomidine, sulphadoxine, fanasil, sulphormethoxine, sulfadoxin, 4-amino-n-5,6-dimethoxypyrimidin-4-yl benzenesulfonamide, sulfadoxina, sulfadoxinum, fanzil
PubChem CID 17134
ChEBI CHEBI:9329
IUPAC Name 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzene-1-sulfonamide
SMILES COC1=NC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1OC

Specifications

Packaging Plastic bottle
Quantity 100 g
Beilstein 625453
Solubility Information Soluble in ethanol & ammonium hydroxide (9:1) at 20mg/ml
Formula Weight 310.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sulfadoxine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.