missing translation for 'onlineSavingsMsg'
Learn More

Sulfaguanidine, 98%, Thermo Scientific Chemicals

Product Code. 10472298 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Product Code. Quantity Packaging unitSize
10472298 500 g Glass bottle 500g
10729808 25 g Glass bottle 25g
10165717 100 g Glass bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 10472298

Brand: Thermo Scientific Acros 431305000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-67-0
Molecular Formula C7H10N4O2S
Molecular Weight (g/mol) 214.25
MDL Number MFCD00038136
InChI Key BRBKOPJOKNSWSG-UHFFFAOYSA-N
Synonym sulfaguanidine, sulphaguanidine, sulfaguanidin, guanicil, sulfaguine, aterian, sulfanilguanidine, sulfanilylguanidine, sulfoguanidine, abiguanil
PubChem CID 5324
IUPAC Name 2-(4-aminophenyl)sulfonylguanidine
SMILES C1=CC(=CC=C1N)S(=O)(=O)N=C(N)N

Specifications

Melting Point 190.5°C to 192.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 97.5% min. (HPLC)
Merck Index 15,904
Formula Weight 214.25
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Sulfaguanidine
Show More Show Less
compliance-icons
Product Identifier
  • Sulfaguanidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.