missing translation for 'onlineSavingsMsg'
Learn More

Sulfaguanidine, TRC

Product Code. 30137603
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137603 10 g 10g
30134700 50 mg 50mg
17914705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137603 Supplier Toronto Research Chemicals Supplier No. TRCS68898510G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-67-0
Molecular Formula C7 H10 N4 O2 S
Molecular Weight (g/mol) 214.24
Synonym (4-Aminophenylsulfonyl)guanidine, Sulfaguanidine, Sulfadimidine Imp. C (Pharmeuropa), N1-(Diaminomethylene)sulfanilamide, 1-Sulfanilylguanidine, 1-[(p-Aminophenyl)sulfonyl]guanidine, 4-Amino-N-(aminoiminomethyl)benzenesulfonamide, 4-Aminobenzenesulfonylguanidine, 4-Aminophenylsulfonylguanidine, A 307, Abiguanil, Aterian, Diacta, Emerin (pharmaceutical), Ganidan, Guamide, Guanicil, Guanidan, N-Guanylsulfanilamide, N1-Amidinosulfanilamide, N1-Guanidylsulfanilamide, N1-Guanylsulfanilamide, NSC 14041, Orgaguanidon, RP 2275, Resulfon, Ruocid, Shigatox, Suganyl, Sulfaguanidin, Sulfaguanil, Sulfaguine, Sulfanilguanidine, Sulfanilylguanidine, Sulfentidine, Sulfoguanidine, Sulfoguanil, Sulfoguanyl, Sulfoguenil, Sulgin, Sulphaguanidine, [(p-Aminophenyl)sulfonyl]guanidine, p-Aminobenzenesulfoguanidide, p-Aminobenzenesulfonylguanidine
IUPAC Name 1-(4-aminophenyl)sulfonylguanidine
SMILES NC(=N)NS(=O)(=O)c1ccc(N)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H10N4O2S/c8-5-1-3-6(4-2-5)14(12,13)11-7(9)10/h1-4H,8H2,(H4,9,10,11)
Formula Weight 214.0524
Percent Purity >95%
Chemical Name or Material Sulfaguanidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.