missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethoxazole-d4 (benzene-d4), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Sulfamethoxazole-d4 |
| CAS | 1020719-86-1 |
| Molecular Formula | C10 2H4 H7 N3 O3 S |
| Quantity | 0.05 g |
| Synonym | Sulfamethoxazole D4 (benzene D4) |
| InChI Formula | InChI=1 S/C10H11N3O3S/c1-7-6-10(12-16-7)13-17(14,15)9-4-2-8(11)3-5-9/h2-6 H,11H2,1H3,(H,12,13)/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1N)S(=O)(=O)Nc2cc(C)on2 |
| IUPAC Name | 4-amino-2,3,5,6-tetradeuterio-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide |
| Molecular Weight (g/mol) | 257.3 |
| Formula Weight | 257.0772 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction