missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic acid, ≥99.5% (alkalimetric), Solstice

Product Code. 15660860
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
4 x 5 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
5kg
Pack of 4
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15660860 1 kg 1kg
15670860 2.5 kg 2.5kg
15600870 5 kg 5kg
15610870 6 x 2.5 kg Pack of 6
15690860 4 x 5 kg Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15660860 Supplier Fluka Supplier No. 074011KG
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

≥99.5% (alkalimetric)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.088
MDL Number MFCD00011603
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

Residue after Ignition ≤0.02%
Quantity 1 kg
Linear Formula NH2SO3H
UN Number 2967
Formula Weight 97.09g/mol
Percent Purity ≥99.5% (alkalimetric)
Chemical Name or Material Sulfamic Acid
compliance-icons
Product Identifier
  • Sulfamic acid
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sulphamidic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.