missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamonomethoxine-d3 (methoxy-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sulfamonomethoxine-d3 |
| Molecular Formula | C11 D3 H9 N4 O3 S |
| Quantity | 0.005 g |
| Synonym | Sulfamonomethoxine D3 (Methoxy D3) |
| InChI Formula | InChI=1 S/C11H12N4O3S/c1-18-11-6-10(13-7-14-11)15-19(16,17)9-4-2-8(12)3-5-9/h2-7 H,12H2,1H3,(H,13,14,15)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1cc(NS(=O)(=O)c2ccc(N)cc2)ncn1 |
| IUPAC Name | 4-amino-N-[6-(trideuteriomethoxy)pyrimidin-4-yl]benzenesulfonamide |
| Molecular Weight (g/mol) | 283.322 |
| Formula Weight | 283.082 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction