missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide, 100.26%, MP Biomedicals™

Product Code. 11473361 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473361 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473361 Supplier MP Biomedicals Supplier No. 0210298890
  • ONLINE EXCLUSIVE PRICE
    182.00 €
    172.20 € / 500g
    Save 9.80 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 63-74-1
Color White
Density 1.08g/mL
Assay Percent Range 1.0026
Molecular Formula C6H8N2O2S
Quantity 500 g
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)S(=O)(=O)N
IUPAC Name 4-aminobenzenesulfonamide
Molecular Weight (g/mol) 172.202
PubChem CID 5333
ChEBI CHEBI:45373
Formula Weight 172.2
Physical Form Crystalline Powder
Chemical Name or Material Sulfanilamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.