missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide, 98%

Product Code. 11489893
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11489893

Brand: Thermo Scientific Alfa Aesar A13001.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63-74-1 | C6H8N2O2S | 172.202 g/mol

Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections. It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. It serves as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfonamide antibacterial Sulfanilamide is an antibacterial and used in the treatment of vaginal yeast infections. It is a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. It serves as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution followed by removal of the sulfonamide blocking group by desulfonation with sulfuric acid.

Solubility
Soluble in water, acetone, ethanol, glycerol, propylene glycol and hydrochloric acid. Insoluble in chloroform, ether, benzene and petroleum ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-74-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00007939
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N

Specifications

Melting Point 163°C to 168°C
Density 1.08
Quantity 2500 g
Beilstein 511852
Merck Index 14,8925
Solubility Information Soluble in water,acetone,ethanol,glycerol,propylene glycol and hydrochloric acid. Insoluble in chloroform,ether,benzene and petroleum ether.
Formula Weight 172.21
Percent Purity 98%
Chemical Name or Material Sulfanilamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.