missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide, MP Biomedicals™

Product Code. 11463361 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463361 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11463361 Supplier MP Biomedicals Supplier No. 0210298880
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as antibacterial agent and to treat vaginal fungal infections. Sulfanilamide, MP Biomedicals is a sulfonamide antibiotic that blocks the synthesis of folic acid by inhibiting the enzyme dihydropteroate synthase. Effective against gram-negative bacter

Sulfanilamide is a molecule containing the sulfonamide functional group attached to an aniline. As a sulfonamide antibiotic, it functions by competitively inhibiting enzymatic reactions (i.e., acting as a substrate analogue) involving para-aminobenzoic acid (PABA). PABA is needed in enzymatic reactions that produce folic acid, which acts as a coenzyme in the synthesis of amino acids. By inhibiting dihydropteroate synthase, it blocks the synthesis of folic acid.

Specifications

CAS 63-74-1
Absorbance λ max (water): 255–259nm
Melting Point 160°C to 170°C
Density 1.08g/mL (Lit.)
Molecular Formula C6H8N2O2S
Quantity 100 g
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
Solubility Information Soluble in 1g dissolves in 37mL alcohol, 5mL acetone; glycerol, ether, benzene, petroleum ether.
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)S(=O)(=O)N
IUPAC Name 4-aminobenzenesulfonamide
Molecular Weight (g/mol) 172.202
PubChem CID 5333
ChEBI CHEBI:45373
Formula Weight 172.2
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.