missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilic Acid, 99+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10439380 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10439380 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10439380 Supplier Fisher Chemical Supplier No. S/8680/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 121-57-3
Melting Point 365°C
Color Blue
Packaging Plastic powder jar
Molecular Formula C6H7NO3S
MDL Number 7886
Quantity 500 g
Synonym sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
InChI Key HVBSAKJJOYLTQU-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)S(=O)(=O)O
IUPAC Name 4-aminobenzenesulfonic acid
Molecular Weight (g/mol) 173.186
PubChem CID 8479
ChEBI CHEBI:27500
Formula Weight 173.19g/mol
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Sulfanilic acid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.