missing translation for 'onlineSavingsMsg'
Learn More

Sulfaquinoxaline, TRC

Product Code. 17974575
Change view
Click to view available options
Quantity:
100 mg
250 mg
10 g
Unit Size:
100mg
10g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17974575 10 g 10g
17913065 250 mg 250mg
17940855 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17974575 Supplier Toronto Research Chemicals Supplier No. TRCS68903510G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-40-5
Molecular Formula C14 H12 N4 O2 S
Molecular Weight (g/mol) 300.34
Synonym 4-Amino-N-2-quinoxalinyl-benzenesulfonamide, Sulfanilamide, N1-2-quinoxalinyl- (6CI,7CI,8CI), 2-(p-Sulfanilamido)quinoxaline, 2-Sulfanilamidobenzopyrazine, 2-p-Aminobenzenesulfonamidoquinoxaline, 2-p-Aminobenzenesulphonamidoquinoxaline, Avicocid, Compound 3-120, Italquina, Kokozigal, N1-(2-Quinoxalinyl)sulfanilamide, NSC 41805, SQ 40, SQX, Sulfa-Q 20, Sulfabenzpyrazine, Sulfaline, Sulfaquinoxaline, Sulphaquinoxaline, Sulquin, Ursokoxaline, Sulfachinoxalin
IUPAC Name 4-amino-N-quinoxalin-2-ylbenzenesulfonamide
SMILES Nc1ccc(cc1)S(=O)(=O)Nc2cnc3ccccc3n2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H12N4O2S/c15-10-5-7-11(8-6-10)21(19,20)18-14-9-16-12-3-1-2-4-13(12)17-14/h1-9H,15H2,(H,17,18)
Formula Weight 300.0681
Percent Purity >95%
Chemical Name or Material Sulfaquinoxaline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.