missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Sulfate Test Kits

Product Code. 11744774
Click to view available options
Range:
100 to 1000 mg/L
5 to 250 mg/L
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11744774

Brand: WTW™ 250414

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Get accurate test results for your ground water, building materials, and soils with the photometric test kits for measurement of Ammonium NH4.

  • Photometric test kit for measurement of Ammonium NH4
  • Variety of instrumentation for measuring CombiCheck Ammonium (NH4)
  • For euse with PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, Photoflex photometers
  • For ground water, building materials, soils
TRUSTED_SUSTAINABILITY

Specifications

Analytes Indophenol blue (Berthelot reaction)
For Use With (Application) For ground water, building materials, soils
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
Includes Certificates
No. of Tests 25
Range 5 to 250 mg/L
Sufficient For 25 Tests
Test Method Photometric
Type CombiCheck Ammonium (NH4) cell test kit
compliance-icons compliance-icons
Product Identifier
  • Sulfate Cell Test Method: photometric 5 - 250 mg/l SO
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Propanol, hydrochloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.