missing translation for 'onlineSavingsMsg'
Learn More

Sulfisoxazole, 99%, Thermo Scientific Chemicals

Product Code. 11946871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11946871 100 g 100g
11936871 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11946871 Supplier Thermo Scientific Acros Supplier No. 439671000
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-69-5
Molecular Formula C11H13N3O3S
Molecular Weight (g/mol) 267.3
InChI Key NHUHCSRWZMLRLA-UHFFFAOYSA-N
Synonym sulfisoxazole, sulfafurazole, sulphafurazole, sulfisoxazol, sulfafurazol, sulfaisoxazole, sulfofurazole, sulfisoxasole, sulphaisoxazole, sulfadimethylisoxazole
PubChem CID 5344
ChEBI CHEBI:102484
IUPAC Name 4-amino-N-(3,4-dimethyl-1,2-oxazol-5-yl)benzenesulfonamide
SMILES CC1=C(ON=C1C)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 194°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Merck Index 15,9082
Solubility Information Solubility in water: 0.13mg/mL (25°C). Other solubilities: soluble in alcohol
Formula Weight 267.3
Percent Purity 99%
Chemical Name or Material Sulfisoxazole
compliance-icons
Product Identifier
  • Sulfisoxazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.