missing translation for 'onlineSavingsMsg'
Learn More

Sulfo Rhodamine Methanethiosulfonate, TRC

Product Code. 30425130
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425130 1 mg 1mg
30419596 10 mg -
30423606 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425130 Supplier Toronto Research Chemicals Supplier No. TRCS6991501MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 386229-71-6
Molecular Formula C30 H37 N3 O8 S4
Molecular Weight (g/mol) 695.89
Synonym Xanthylium, 3,6-bis(diethylamino)-9-[4-[[[2-[(methylsulfonyl)thio]ethyl]amino]sulfonyl]-2-sulfophenyl]-, inner salt, MTS-rhodamine, Sulfo Rhodamine Methanethiosulfonate
IUPAC Name 2-[3,6-bis(diethylamino)xanthen-10-ium-9-yl]-5-(2-methylsulfonylsulfanylethylsulfamoyl)benzenesulfonate
SMILES CCN(CC)c1ccc2c(c3ccc(cc3S(=O)(=O)[O-])S(=O)(=O)NCCSS(=O)(=O)C)c4ccc(cc4[o+]c2c1)N(CC)CC

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C30H37N3O8S4/c1-6-32(7-2)21-10-13-24-27(18-21)41-28-19-22(33(8-3)9-4)11-14-25(28)30(24)26-15-12-23(20-29(26)45(38,39)40)44(36,37)31-16-17-42-43(5,34)35/h10-15,18-20,31H,6-9,16-17H2,1-5H3
Formula Weight 695.1463
Chemical Name or Material Sulforhodamine Methanethiosulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.