missing translation for 'onlineSavingsMsg'
Learn More

Sulfobromophthalein Sodium Salt Crystalline MP Biomedicals

Product Code. 11495141 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495141 5 g 5g
11405151 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495141 Supplier MP Biomedicals Supplier No. 0215672605
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 71-67-0
Molecular Formula C20H8Br4Na2O10S2
Molecular Weight (g/mol) 837.99
MDL Number MFCD00150017
InChI Key GHAFORRTMVIXHS-UHFFFAOYSA-L
Synonym bromosulfophthalein, sulfobromophthalein, 3h bsp, 3h-bsp, d00ady
PubChem CID 102371197
ChEBI CHEBI:63827
SMILES [Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C(=C1)S([O-])(=O)=O

Specifications

Quantity 5 g
IUPAC Name disodium;2-sulfo-4-[4,5,6,7-tetrabromo-1-(4-oxido-3-sulfophenyl)-3-oxo-2-benzofuran-1-yl]phenolate
Formula Weight 838
Physical Form Crystalline
Chemical Name or Material Sulfobromophthalein Sodium Salt
compliance-icons compliance-icons
Product Identifier
  • Sulfobromophthalein Sodium Salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: SulfobromophthaleinSodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.