missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulfobromophthalein sodium salt hydrate

Product Code. 11320558
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11320558 25 g 25g
11300558 1 g 1g
11310558 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11320558 Supplier Thermo Scientific Chemicals Supplier No. 043878.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 71-67-0 | C20H8Br4Na2O10S2 | 837.99 g/mol

Sulfobromophthalein sodium salt hydrate is a reagent for the quantitative determination of proteins, a dye for liver diagnostics in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-67-0
Molecular Formula C20H8Br4Na2O10S2
Molecular Weight (g/mol) 837.99
MDL Number MFCD00150017
InChI Key GHAFORRTMVIXHS-UHFFFAOYSA-L
Synonym bromosulfophthalein, sulfobromophthalein, 3h bsp, 3h-bsp, d00ady
PubChem CID 102371197
ChEBI CHEBI:63827
IUPAC Name disodium;2-sulfo-4-[4,5,6,7-tetrabromo-1-(4-oxido-3-sulfophenyl)-3-oxo-2-benzofuran-1-yl]phenolate
SMILES [Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C(=C1)S([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Sensitivity Moisture sensitive
Merck Index 14,8954
Solubility Information Soluble in water.
Formula Weight 837.98 (Anhydrous)
Physical Form Powder
Chemical Name or Material Sulfobromophthalein sodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.