missing translation for 'onlineSavingsMsg'
Learn More

Sulfobromophthalein Sodium Salt, MP Biomedicals™

Product Code. 11485141 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485141 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11485141 Supplier MP Biomedicals Supplier No. 0215672601
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used as a substrate for multidrug resistance associated protein 2 (Mrp2). Sulfobromophthalein Sodium Salt, MP Biomedicals is a dye with a high affinity for organic anion transporting polypeptides (OATPs). BSP is used in diagnosis of hepatic disorders and

BSP is transported into hepatocytes by OATPs and, after conjugation to glutathione, is excreted into bile by Mrp2.3 It was found to inhibit the aldo-keto reductase ARK1C20.

TRUSTED_SUSTAINABILITY

Specifications

CAS 71-67-0
Molecular Formula C20H8Br4Na2O10S2
MDL Number MFCD00150017
Quantity 1 g
Synonym bromosulfophthalein, sulfobromophthalein, 3h bsp, 3h-bsp, d00ady
Solubility Information Soluble in water (20mg/mL, clear, colorless solution). Insoluble in ethanol or acetone.
InChI Key GHAFORRTMVIXHS-UHFFFAOYSA-L
SMILES [Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C(=C1)S([O-])(=O)=O
IUPAC Name disodium 2-hydroxy-5-[4,5,6,7-tetrabromo-1-(4-hydroxy-3-sulfonatophenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]benzene-1-sulfonate
Molecular Weight (g/mol) 837.99
PubChem CID 102371197
ChEBI CHEBI:63827
Formula Weight 838
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.