missing translation for 'onlineSavingsMsg'
Learn More

Sulfolane, 80% w/w aq. soln.

Product Code. 15425237
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425237 500 mL 500mL
15415237 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425237 Supplier Thermo Scientific Alfa Aesar Supplier No. H32505.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 126-33-0 | C4H8O2S | 120.17 g/mol

Sulfolane is used in the chemical industry as a solvent for extractive distillation and chemical reactions. It is used to separate aromatic hydrocarbons (benzene, toluene and xylenes), to separate n-propyl alcohol and sec-butyl alcohol. It is also used to purify natural gas streams and for fractionation of fatty acids into saturated and unsaturated components and as a reaction solvent for the preparation of aromatic sulfonic acids, pyridines, isocyanates and pharmaceuticals. It can also be involved in the halogen exchange process and polymerization process. It is used as a plasticizer and curing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfolane is used in the chemical industry as a solvent for extractive distillation and chemical reactions. It is used to separate aromatic hydrocarbons (benzene, toluene and xylenes), to separate n-propyl alcohol and sec-butyl alcohol. It is also used to purify natural gas streams and for fractionation of fatty acids into saturated and unsaturated components and as a reaction solvent for the preparation of aromatic sulfonic acids, pyridines, isocyanates and pharmaceuticals. It can also be involved in the halogen exchange process and polymerization process. It is used as a plasticizer and curing agent.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-33-0
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00005484
InChI Key HXJUTPCZVOIRIF-UHFFFAOYSA-N
Synonym sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
PubChem CID 31347
ChEBI CHEBI:74794
IUPAC Name thiolane 1,1-dioxide
SMILES O=S1(=O)CCCC1

Specifications

Melting Point 0°C to 5°C
Density 1.212
Odor Weak, Characteristic
Quantity 500 mL
Beilstein 107765
Merck Index 14,8955
Solubility Information Fully miscible in water.
Formula Weight 120.17
Concentration or Composition (by Analyte or Components) 80% w/w aq. soln.
Chemical Name or Material Sulfolane
compliance-icons compliance-icons
Product Identifier
  • Sulfolane, 80% w/w aq. soln.
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tetramethylene sulfone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.