missing translation for 'onlineSavingsMsg'
Learn More

Sulfolane, TRC

Product Code. 30427545
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427545 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427545 Supplier Toronto Research Chemicals Supplier No. TRCS689220500G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfolan
CAS 126-33-0
Molecular Formula C4 H8 O2 S
Quantity 500 g
Synonym Sulfolan, Thiophene','tetrahydro-','1,1-dioxide, 1,1-Dioxothiolan, 1λ6-Thiolane-1,1-dione, Bondelane A, Bondolane A, Cyclic tetramethylene sulfone, Cyclotetramethylene sulfone, NSC 46443, Sulfolan, Sulfolane, Sulfolane SG, Sulpholane, Tetrahydrothiophene 1,1-dioxide, Tetrahydrothiophene S,S-dioxide, Tetrahydrothiophene dioxide, Tetramethylene sulfone, Thiacyclopentane dioxide, Thiolane 1,1-dioxide, Thiophane 1,1-dioxide, Thiophane dioxide
InChI Formula InChI=1S/C4H8O2S/c5-7(6)3-1-2-4-7/h1-4H2
SMILES O=S1(=O)CCCC1
IUPAC Name thiolane 1,1-dioxide
Molecular Weight (g/mol) 120.17
Formula Weight 120.0245
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.