missing translation for 'onlineSavingsMsg'
Learn More

Sulfonamide, MP Biomedicals

Product Code. 11453361 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453361 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453361 Supplier MP Biomedicals Supplier No. 0210298825
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 63-74-1
Molecular Formula C6H8N2O2S
Quantity 25 g
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)S(=O)(=O)N
IUPAC Name 4-aminobenzenesulfonamide
Molecular Weight (g/mol) 172.202
PubChem CID 5333
ChEBI CHEBI:45373
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.