missing translation for 'onlineSavingsMsg'
Learn More

Sulforhodamine 101 acid chloride

Product Code. 15403339
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15403339 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15403339 Supplier Thermo Scientific Alfa Aesar Supplier No. J60581.MA
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82354-19-6 | C31H29ClN2O6S2 | 625.15 g/mol

Fluorescent labeling compound

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 82354-19-6
Sensitivity Light and moisture sensitive
Molecular Formula C31H29ClN2O6S2
MDL Number MFCD00012406
Beilstein 8667894
Synonym Texas Red™; Sulforhodamine 101 sulfonyl chloride
Color Red
Quantity 10 mg
InChI Key MPLHNVLQVRSVEE-UHFFFAOYSA-N
Physical Form Powder
SMILES [O-]S(=O)(=O)C1=C(C=CC(=C1)S(Cl)(=O)=O)C1=C2C=C3CCC[N+]4=C3C(CCC4)=C2OC2=C3CCCN4CCCC(C=C12)=C34
IUPAC Name 16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3-oxa-9λ⁵,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,4,9(28),13,15,17,19(27)-heptaen-9-ylium
Molecular Weight (g/mol) 625.15
PubChem CID 452705
ChEBI CHEBI:51247
Formula Weight 625.15
Chemical Name or Material Sulforhodamine 101 acid chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulforhodamine 101 acid chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Sulforhodamine 101 acid chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.