missing translation for 'onlineSavingsMsg'
Learn More

Sulforhodamine B, MP Biomedicals™

Product Code. 11472752 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g

Product Code. 11472752

Brand: MP Biomedicals 0215247901

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 3520-42-1
Molecular Formula C29H33N2NaO7S2
Molecular Weight (g/mol) 608.70
MDL Number MFCD00010180
InChI Key XJENLUNLXRJLEZ-UHFFFAOYSA-M
Synonym Acid Red 52, Phloxinrhodamine
PubChem CID 131852807
IUPAC Name sodium 3,6-bis(diethylamino)-9-(2,4-disulfonatophenyl)-2,7-dimethyl-10λ⁴-xanthen-10-ylium
SMILES [Na+].CCN(CC)C1=CC2=[O+]C3=CC(N(CC)CC)=C(C)C=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C2C=C1C

Specifications

Color Brown-Purple
Quantity 1 g
Formula Weight 580.66
Physical Form Powder
Chemical Name or Material Sulforhodamine B
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.