missing translation for 'onlineSavingsMsg'
Learn More

Sulfosuccinimidyl 3-[[2-(Biotinamido)ethyl] dithio]propionate Sodium Salt (Technical Grade), TRC

Product Code. 30427979
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427979 500 mg 500mg
30422909 1 g -
30425750 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427979 Supplier Toronto Research Chemicals Supplier No. TRCS699450500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 325143-98-4
Molecular Formula C19 H27 N4 O9 S4 . Na
Molecular Weight (g/mol) 606.69
IUPAC Name sodium;1-[3-[2-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]ethyldisulfanyl]propanoyloxy]-2,5-dioxopyrrolidine-3-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1CC(=O)N(OC(=O)CCSSCCNC(=O)CCCC[C@@H]2SC[C@@H]3NC(=O)N[C@H]23)C1=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H28N4O9S4.Na/c24-14(4-2-1-3-12-17-11(10-33-12)21-19(28)22-17)20-6-8-35-34-7-5-16(26)32-23-15(25)9-13(18(23)27)36(29,30)31;/h11-13,17H,1-10H2,(H,20,24)(H2,21,22,28)(H,29,30,31);/q;+1/p-1/t11-,12-,13?,17-;/m0./s1
Formula Weight 606.0559
Chemical Name or Material Sulfosuccinimidyl 3-[[2-(Biotinamido)ethyl] dithio]propionat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.