missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Dioxide Solution, Honeywell Fluka™

Product Code. 15652690 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15652690 100 g 100g
15672690 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15652690 Supplier Honeywell-Fluka Supplier No. 247626100G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7782-99-2
Molecular Formula H2O3S
Molecular Weight (g/mol) 82.073
MDL Number MFCD00011335
InChI Key LSNNMFCWUKXFEE-UHFFFAOYSA-N
Synonym sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid
PubChem CID 1100
ChEBI CHEBI:48854
IUPAC Name sulfurous acid
SMILES OS(=O)O

Specifications

Quantity 100 g
Linear Formula H2SO3
UN Number 1833
Formula Weight 82.08g/mol
Percent Purity ≥6%
Grade ACS Reagent
Residue after Ignition ≤0.005%
Density 1.03g/mL at 25°C; Vapor Density: 2.3 (vs air)
Chemical Name or Material Sulfur Dioxide Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.