missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, Extra Pure, Precipitated, SLR, Fisher Chemical

Product Code. 10020380 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10020380 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10020380 Supplier Fisher Chemical Supplier No. S/8990/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 445°C
Melting Point 113°C
CAS 7704-34-9
Color Yellow
Physical Form Solid
Quantity 500 g
Molecular Formula S
MDL Number MFCD00085316
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES [S]
IUPAC Name sulfur
Molecular Weight (g/mol) 32.06
PubChem CID 5362487
ChEBI CHEBI:26833
Formula Weight 32.06g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Sulfur precipitated
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.