missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, purum p.a., ≥99.5% (T), Honeywell™ Fluka™

Product Code. 15627320 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15627320 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15627320 Supplier Honeywell Research Chemicals Supplier No. 846831KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥99.5% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7704-34-9
Quantity 1 kg
Molecular Formula S
MDL Number MFCD00085316
UN Number UN1350
Flash Point 207°C (404.6°F)
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES [S]
IUPAC Name sulfur
Molecular Weight (g/mol) 32.06
PubChem CID 5362487
ChEBI CHEBI:26833
Formula Weight 32.07g/mol
Percent Purity ≥99.5% (T)
Ignition Point 450°F
Packaging Poly bottle
Vapor Pressure 10mmHg (246°C), 1mmHg (183.8°C)
Density Vapor Density: 8.9 (vs air)
Chemical Name or Material Sulfur
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.