missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, Reagent grade, Purified by refining, Honeywell™

Product Code. 15631980 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15631980 2 kg Poly Bottle 2kg
15641980 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15631980 Supplier Honeywell Research Chemicals Supplier No. 2151982KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, powder, purified by refining, -100 mesh particle size

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Boiling Point 445°C
Melting Point 120°C
Color Yellow
Physical Form Powder
Quantity 2 kg
UN Number UN1350
Flash Point 207°C (404.6°F)
Formula Weight 32.07g/mol
Grade Reagent
Ignition Point 450°F
Packaging Poly Bottle
Vapor Pressure 1mmHg (183.8°C), 10mmHg (246°C)
Density 2.07g/cu.cm; Vapor Density: 8.9 (vs air)
Chemical Name or Material Sulfur, Purified by refining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.