missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, Technical, Roll, Fisher Chemical

Product Code. 10549580 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10549580 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10549580 Supplier Fisher Chemical Supplier No. S/8970/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point 445°C
Melting Point 113°C
CAS 7704-34-9
Color Yellow
Physical Form Solid
Quantity 1 kg
Molecular Formula S
MDL Number MFCD00085316
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES [S]
IUPAC Name sulfur
Molecular Weight (g/mol) 32.06
PubChem CID 5362487
ChEBI CHEBI:26833
Formula Weight 32.06g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.