missing translation for 'onlineSavingsMsg'
Learn More

Sulfur trioxide-pyridine complex, 98%, active SO3 ca 48-50%

Product Code. 11484863
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1000 g
Unit Size:
1000g
10g
250g
50g
Product Code. Quantity unitSize
11484863 250 g 250g
11423960 10 g 10g
11474863 50 g 50g
11464863 1000 g 1000g
4 options
This item is not returnable. View return policy

Product Code. 11484863

Brand: Thermo Scientific Alfa Aesar A12202.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26412-87-3 | C5H5NO3S | 159.16 g/mol

Sulfur trioxide-pyridine complex is used for sulfations of alcohols, sulfonations, deoxygenations and other reductions, oxidations with dimethyl sulfoxide. It is also used as a source of sulfur trioxide especially in the preparation of sulfate esters. Further, it is used as an activating electrophile in a Parikh-Doering oxidation. In addition to this, it is also used in the preparation and anticoagulation test of sulfated silk fibroin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfur trioxide-pyridine complex is used for sulfations of alcohols, sulfonations, deoxygenations and other reductions, oxidations with dimethyl sulfoxide. It is also used as a source of sulfur trioxide especially in the preparation of sulfate esters. Further, it is used as an activating electrophile in a Parikh-Doering oxidation. In addition to this, it is also used in the preparation and anticoagulation test of sulfated silk fibroin.

Solubility
Soluble in polar organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26412-87-3
Molecular Formula C5H5NO3S
Molecular Weight (g/mol) 159.16
MDL Number MFCD00012437
InChI Key UDYFLDICVHJSOY-UHFFFAOYSA-N
Synonym pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide
PubChem CID 168533
SMILES O=S(=O)=O.C1=CC=NC=C1

Specifications

Melting Point ∼155°C to 165°C
Quantity 250 g
UN Number UN3261
Beilstein 3704116
Sensitivity Moisture sensitive
Solubility Information Soluble in polar organic solvents.
IUPAC Name pyridine;sulfur trioxide
Formula Weight 159.16
Percent Purity 98%
Chemical Name or Material Sulfur trioxide-pyridine complex, Active SO3 ≈48 to 50%
compliance-icons
Product Identifier
  • Sulphur trioxide-pyridine complex
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sulphur trioxide-pyridine complex

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.