missing translation for 'onlineSavingsMsg'
Learn More

Sulfur trioxide trimethylamine complex, 95%

Product Code. 10687124 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10687124 100 g Glass bottle 100g
10055823 25 g Glass bottle 25g
10484765 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10687124 Supplier Thermo Scientific Acros Supplier No. 159351000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3162-58-1 | C3H9NO3S | 139.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3162-58-1
Molecular Formula C3H9NO3S
Molecular Weight (g/mol) 139.17
MDL Number MFCD00012421
InChI Key DXASQZJWWGZNSF-UHFFFAOYSA-N
Synonym sulfur trioxide trimethylamine complex, sulfur trioxide trimethylamine, sulfur trioxide-trimethylamine complex, sulphur trioxide trimethylamine 1:1, sulfur trioxide-trimethylamine, trimethylamine sulfur trioxide, sulfur trioxide; trimethylamine, 1n1&1&&so3 complex, trimethylamine, compd. with sulfur trioxide 1:1, trimethylamine, compound with sulphur trioxide
PubChem CID 222852
IUPAC Name N,N-dimethylmethanamine;sulfur trioxide
SMILES CN(C)C.O=S(=O)=O

Specifications

Melting Point 232.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (CH3)3N·SO3
Quantity 100 g
Fieser 01,1128; 08,466
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot water, ethanol
Formula Weight 139.17
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Sulfur trioxide trimethylamine complex
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfur trioxide trimethylamine complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.