missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Trioxide Trimethylamine Complex, TRC

Product Code. 30416418
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416418 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416418 Supplier Toronto Research Chemicals Supplier No. TRCS7894651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfur Trioxide Trimethylamine Complex
CAS 3162-58-1
Molecular Formula C3H9NO3S
Quantity 1 g
Synonym N,N-Dimethyl-methanamine Compd. with Sulfur Trioxide, Trimethylamine Compd. with Sulfur Trioxide, Sulfur Trioxide Compd. with N,N-Dimethylmethanamine, Sulfur Trioxide Compd. with Trimethylamine, NSC 9838, Sulfur Trioxide-trimethylamine, Sulfur Trioxide-trimethylamine Complex, Trimethylamine Compound with Sulfur Trioxide, Trimethylamine Sulfur Trioxide
InChI Formula InChI=1S/C3H9N.O3S/c2*1-4(2)3/h1-3H3;
SMILES CN(C)C.O=S(=O)=O
IUPAC Name N,N-dimethylmethanamine;sulfur trioxide
Molecular Weight (g/mol) 139.17
Formula Weight 139.0303
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.