missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.05M, Chem-Lab

Product Code. 11933293
Change view
Click to view available options
Quantity:
1 L
Packaging:
Polyethylene
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11933293 1 L Polyethylene 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11933293 Supplier Chem-Lab Supplier No. CL0526121000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal for titrations in laboratory applications. Sulfuric Acid, 0.05M, Chem-Lab is a solution of 4.905g H2SO4 per liter of water. It is manufactured according to ASTM D-3228 standards.

Sulfuric acid is a highly corrosive, strong mineral acid that shows different properties depending upon its concentration. It is a pungent-ethereal, colorless to slightly yellow viscous liquid. Sulfuric acid is a diprotic acid that is hygroscopic. Concentrated sulfuric acid has strong dehydrating and oxidizing properties. Dilute sulfuric acid is a good electrolyte.

Specifications

pH <2
Color Colorless
CAS 7664-93-9, 7732-18-5
Composition 4.905g H2SO4 per 1L H2O
Melting Point 0°C
Boiling Point 100°C
Concentration 0.05mol/L
Density 1.01g/mL
Odor Odorless
Packaging Polyethylene
Normality 0.1N (±0.0002, 20°C)
Quantity 1 L
Molecular Formula H2O4S
Viscosity 1 cSt
Physical Form Liquid
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Solubility Information Soluble in Water
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.08g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.