missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid 95-98% d=1.83, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™

Product Code. 10466463 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10466463 1 L HDPE plastic bottle 1 L
10620714 500 mL Plastic Coated Glass bottle 500mL
10707263 2.5 L HDPE plastic bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10466463 Supplier Fisher Chemical Supplier No. S/9231/PB15
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric Acid 95-98% d=1.83, Primar Plus™, for Trace Metal Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 1
Color Colorless
Melting Point 10°C
Boiling Point 290°C
Packaging HDPE plastic bottle
Vapor Pressure 1mmHg 146
Quantity 1 L
Viscosity 21mPa at 25°C
Physical Form Liquid
Formula Weight 98.07g/mol
compliance-icons
Product Identifier
  • Sulfuric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.